missing translation for 'onlineSavingsMsg'
Learn More

Trap 101, Tocris Bioscience™

Potent and selective NOP antagonist; active in vivo

Brand:  Tocris (Bio-Techne) 250850

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13461364

  • £671.97 / 50mg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Potent and selective nociceptin/orphanin FQ (NOP) receptor antagonist (pA2 = 7.75). Displays selectivity for NOP receptors over classical opioid receptors (pKi values are 8.65, 6.60, 6.14 and < 5 for NOP,μ-, κ-, and δ-opioid receptors respectively). Attenuates motor deficits in a rat model of Parkinson's Disease. Active in vivo.
Specifications

Chemical Identifiers

1216621-00-9
434.021
trap 101, trap-101 hydrochloride, 1-1-cyclooctylmethyl-1,2,3,6-tetrahydro-5-hydroxymethyl-4-pyridinyl-3-ethyl-1,3-dihydro-2h-benzimidazol-2-one hydrochloride, 1-1-cyclooctylmethyl-5-hydroxymethyl-1,2,3,6-tetrahydropyridin-4-yl-3-ethyl-1h-benzo d imidazol-2 3h-one hydrochloride, 1-1-cyclooctylmethyl-3-hydroxymethyl-5,6-dihydro-2h-pyridin-4-yl-3-ethyl-1,3-benzodiazol-2-one hydrochloride, trap101-hcl, trap-101 hydrochloride hplc , powder, 1-1-cyclooctylmethyl-1,2,3,6-tetrahydro-5-hydroxymethyl-4-pyridinyl-3-ethyl-1,3-dihydro-2hbenzimidazol-2-one hydrochloride
1-[1-(cyclooctylmethyl)-5-(hydroxymethyl)-3,6-dihydro-2H-pyridin-4-yl]-3-ethylbenzimidazol-2-one;hydrochloride
C24H36ClN3O2
YEVMLMQAMCKVSC-UHFFFAOYSA-N
56972178
CCN1C2=CC=CC=C2N(C1=O)C3=C(CN(CC3)CC4CCCCCCC4)CO.Cl

Specifications

Trap 101
1216621-00-9
trap 101, trap-101 hydrochloride, 1-1-cyclooctylmethyl-1,2,3,6-tetrahydro-5-hydroxymethyl-4-pyridinyl-3-ethyl-1,3-dihydro-2h-benzimidazol-2-one hydrochloride, 1-1-cyclooctylmethyl-5-hydroxymethyl-1,2,3,6-tetrahydropyridin-4-yl-3-ethyl-1h-benzo d imidazol-2 3h-one hydrochloride, 1-1-cyclooctylmethyl-3-hydroxymethyl-5,6-dihydro-2h-pyridin-4-yl-3-ethyl-1,3-benzodiazol-2-one hydrochloride, trap101-hcl, trap-101 hydrochloride hplc , powder, 1-1-cyclooctylmethyl-1,2,3,6-tetrahydro-5-hydroxymethyl-4-pyridinyl-3-ethyl-1,3-dihydro-2hbenzimidazol-2-one hydrochloride
CCN1C2=CC=CC=C2N(C1=O)C3=C(CN(CC3)CC4CCCCCCC4)CO.Cl
434.021
434.02
50mg
C24H36ClN3O2
YEVMLMQAMCKVSC-UHFFFAOYSA-N
1-[1-(cyclooctylmethyl)-5-(hydroxymethyl)-3,6-dihydro-2H-pyridin-4-yl]-3-ethylbenzimidazol-2-one;hydrochloride
56972178
>98%
Videos