missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ HDPE General Purpose Funnels

Manufactured from HDPE for optimum strength and chemical resistance. Kartell™ HDPE General Purpose Funnels provide safety and precision in filling bottles in the laboratory.

Brand:  Kartell™ 176

Code : NEW

Additional Details : Weight : 0.15100kg

 View more versions of this product

Product Code. 10586001

  • £5.25 / Each


In Stock 11
Add to basket

You may only order up to the available stock. For more details call customer services on 01509 555 500

Description

Description

  • Suitable for a wide range of liquids
  • Upstand on rim reduces potential overflow
  • Outer ribbing eliminates back pressure 
  • Hanging eye for storage
Specifications

Specifications

35mm
Suitable for a wide range of liquids
2750mL
220mm
81mm
HDPE (High-density Polyethylene)
Videos