missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ HDPE General Purpose Funnels

Manufactured from HDPE for optimum strength and chemical resistance. Kartell™ HDPE General Purpose Funnels provide safety and precision in filling bottles in the laboratory.

Brand:  Kartell™ 4171

Code : NEW

Additional Details : Weight : 0.01128kg

 View more versions of this product

Product Code. 10336111

  • £1.10 / Each


In Stock 12
Add to basket

Description

Description

  • Suitable for a wide range of liquids
  • Upstand on rim reduces potential overflow
  • Outer ribbing eliminates back pressure 
  • Hanging eye for storage
Specifications

Specifications

13mL
Suitable for a wide range of liquids
100mL
80mm
27mm
HDPE (High-density Polyethylene)
Videos