missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ VITsafe™ 1000mL Wide-Mouth Wash Bottles

Imprinted with durable safety symbols, and other important information for optimum laboratory safety. Vitlab™ VITsafe™ Wide-Mouth Wash Bottles include color coded vent-cap that supports easy visual identification of the media.

Brand:  VITLAB™ 1453829

Code : A0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10287802

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Manufactured from PE-LD (or PP for Acetone and MEK), spray tube from PP
  • Complies in accordance to Directive (EC) No. 1272/2008 (GHS)
  • Imprinted with:
    • Material name in German, English, French and Spanish
    • Chemical formula, CAS No, hazard pictogram, signal word
    • Risk Phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
  • Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube
  • The VENT-CAP design oprevents almost all static overpressure minimizing leakage or dripping
Specifications

Specifications

Polypropylene
Vented
1000 mL
Polypropylene
Directive (EC) No. 1272/2008 (GHS)
Wide
226 mm
91 mm
Imprint: Acetone
Red
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers