missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Translucent Polypropylene Volumetric Flasks with Stopper

Brand:  Kartell™ 1484

Code : NEW

Additional Details : Weight : 0.00800kg

 View more versions of this product

Product Code. 11328533

  • £6.30 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

Yes
Translucent
25mL
121mm
Stopper
36mm
PP (Polypropylene)
Stopper
Videos