missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Translucent Polypropylene Volumetric Flasks with Stopper

Brand:  Kartell™ 1487

Code : 02

Additional Details : Weight : 1.50000kg

 View more versions of this product

Product Code. 11358533

  • £22.55 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

Yes
Translucent
250mL
225mm
Stopper
82mm
PP (Polypropylene)
Stopper
Videos
Alternative Products