missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Translucent Polypropylene Volumetric Flasks with Screw Cap

Brand:  Kartell™ 1434

Code : 03

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11371225

  • £11.40 / Each


In Stock 5
Add to basket

Description

Description

These flasks create no meniscus so final readings are easy. Thick walls eliminate distortion and thin neck make final reading precise. Chemically non-absorbent. Autoclavable.
Specifications

Specifications

Yes
Translucent
500mL
265mm
10Pack
Screw cap
100mm
PP (Polypropylene)
Screw Caps
Videos
Documentation