missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Translucent Polypropylene Volumetric Flasks with Screw Cap

Brand:  Kartell™ 1430

UNSPSC : 41121804

Code : 03

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11361225

  • £21.75 / Each


In Stock 11
Add to basket

Specifications

Description

These flasks create no meniscus so final readings are easy. Thick walls eliminate distortion and thin neck make final reading precise. Chemically non-absorbent. Autoclavable.
Description & Specifications

Specifications

Yes
Translucent
121mm
25mL
Screw cap
36mm
Screw Caps
PP (Polypropylene)
Documentation