missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Translucent Polypropylene Volumetric Flasks with Screw Cap

Brand:  Kartell™ 1432

UNSPSC : 41121804

Code : 03

Additional Details : Weight : 0.01000kg

Product Code. 11331225

  • £26.60 / Each


In Stock 14
Add to basket

Specifications

Description

These flasks create no meniscus so final readings are easy. Thick walls eliminate distortion and thin neck make final reading precise. Chemically non-absorbent. Autoclavable.
Description & Specifications

Specifications

Yes
Translucent
174mm
100mL
Screw cap
58mm
Screw Caps
PP (Polypropylene)
Documentation