Translucent Crucible Lids
Overview
Brand: Yorlab
Manufacturer Part Number: 10444/28
UNSPSC: 41122201
Code: 03
Additional Details:
Additional Details: Weight: 1.00000kg
Product Code. 12877733
Quantity Price
1 £ 63.56 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description and Specification
Specification
Material Silica
Product Type Crucible Lid
Color Translucent
Diameter (Metric) 82mm