missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Rat Estrogen receptor siRNA
Blocks transcription and translation of Estrogen receptor from Rat.
£227.54 - £336.64
Specifications
Accession Number | NP_036821 |
---|---|
Cell Lines Tested | Rat Primary Osteoblasts. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | Esr1 |
Gene ID (Entrez) | 24890 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16059171
|
enQuireBio™
QX15-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16049171
|
enQuireBio™
QX15-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_036821 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
24890 | |
RNESTROR, ER-alpha, Esr, ESRA, ERA, ERAlpha, ER | |
Lyophilized |
Rat Primary Osteoblasts. | |
Esr1 | |
siRNA | |
Rat |