missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human TLR4 siRNA
Blocks transcription and translation of TLR4 from Human.
£227.54 - £336.64
Specifications
Accession Number | NP_612564 |
---|---|
Cell Lines Tested | HepG2 Human Carcinoma Cells from Liver. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | TLR4 |
Gene ID (Entrez) | 7099 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16099201
|
enQuireBio™
QX30-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089201
|
enQuireBio™
QX30-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_612564 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
7099 | |
TLR-4, Toll-like Receptor 4, CD284 | |
Lyophilized |
HepG2 Human Carcinoma Cells from Liver. | |
TLR4 | |
siRNA | |
Human |