Learn More
Transition Metals, plasma standard solution, Specpure™, Cd, Co, Cr, Cu, Fe, Mn, Ni, V, Zn at 100μg/mL, Thermo Scientific Chemicals
£119.00 - £338.00
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11405260
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
044518.AE
100 mL
£119.00
100mL
View alternative products
Save Up to 
»
Estimated Shipment: 29-04-2024
to see stock.
11415260
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
044518.AP
500 mL
£338.00
500mL
View alternative products
Save Up to 
»
Estimated Shipment: 29-04-2024