missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-4-(3,4-Difluorophenyl)-4'-n-pentylbicyclohexyl, 97%, Thermo Scientific Chemicals

Catalog Number p-7036779
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
118164-51-5
C23H34F2
348.522
MFCD13182310
YDVUSMRUBCJGAV-UHFFFAOYSA-N
trans,trans-4-3,4-difluorophenyl-4'-pentyl-1,1'-bi cyclohexane, trans,trans-4-3,4-difluorophenyl-4'-pentylbicyclohexyl, 4-trans-4-trans-4-pentylcyclohexyl cyclohexyl-1,2-difluorobenzene, benzene, 1,2-difluoro-4-4'-pentyl 1,1'-bicyclohexyl-4-yl, 1,2-difluoro-4-trans,trans-4'-pentyl 1,1'-bicyclohexyl-4-yl benzene, 1,2-difluoro-4-4-4-pentylcyclohexyl cyclohexyl benzene, 4-3,4-difluorophenyl-4'-pentyl-1,1'-bi cyclohexane, trans,trans-4-3,4-difluorophenyl-4'-n-pentylbicyclohexyl, 1r,1's,4r,4'r-4-3,4-difluorophenyl-4'-pentyl-1,1'-bi cyclohexane, trans,trans-4-pentylbicyclohexyl-3,4-difluorobenzene
581521
1,2-difluoro-4-[4-(4-pentylcyclohexyl)cyclohexyl]benzene
CCCCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F
This item is not returnable. View return policy
118164-51-5
C23H34F2
348.522
MFCD13182310
YDVUSMRUBCJGAV-UHFFFAOYSA-N
trans,trans-4-3,4-difluorophenyl-4'-pentyl-1,1'-bi cyclohexane, trans,trans-4-3,4-difluorophenyl-4'-pentylbicyclohexyl, 4-trans-4-trans-4-pentylcyclohexyl cyclohexyl-1,2-difluorobenzene, benzene, 1,2-difluoro-4-4'-pentyl 1,1'-bicyclohexyl-4-yl, 1,2-difluoro-4-trans,trans-4'-pentyl 1,1'-bicyclohexyl-4-yl benzene, 1,2-difluoro-4-4-4-pentylcyclohexyl cyclohexyl benzene, 4-3,4-difluorophenyl-4'-pentyl-1,1'-bi cyclohexane, trans,trans-4-3,4-difluorophenyl-4'-n-pentylbicyclohexyl, 1r,1's,4r,4'r-4-3,4-difluorophenyl-4'-pentyl-1,1'-bi cyclohexane, trans,trans-4-pentylbicyclohexyl-3,4-difluorobenzene
581521
1,2-difluoro-4-[4-(4-pentylcyclohexyl)cyclohexyl]benzene
CCCCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F

It is used as flavor and fragrance intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as flavor and fragrance intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Slightly soluble in water.

Notes
Reacts with strong oxidizing agents. Keep at room temperature.
TRUSTED_SUSTAINABILITY
Melting Point 45°C to 47°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 348.52
Percent Purity 97%
Chemical Name or Material trans,trans-4-(3,4-Difluorophenyl)-4'-n-pentylbicyclohexyl

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'