missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans,trans-2,4-Hexadien-1-ol, 99%, stabilized, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 30.5°C to 31.5°C |
Color | Colorless to Yellow |
Density | 0.8700g/mL |
Boiling Point | 80.0°C (12.0 mmHg) |
Flash Point | 72°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass Bottle |
Linear Formula | CH3CH=CHCH=CHCH2OH |
Refractive Index | 1.4990 to 1.5010 |
Show More |
Safety and Handling
Product Identifier
- trans,trans-2,4-Hexadien-1-ol
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H311-Toxic in contact with skin.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'