missing translation for 'onlineSavingsMsg'
Learn More

trans-beta-Nitrostyrene, 97+%, Thermo Scientific Chemicals

Catalog Number p-3779344
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5153-67-3
C8H7NO2
149.15
MFCD00007402
PIAOLBVUVDXHHL-VOTSOKGWSA-N
trans-beta-nitrostyrene, 2-nitrovinyl benzene, e-2-nitrovinyl benzene, beta-nitrostyrene, 2-nitro-1-phenylethylene, e-beta-nitrostyrene, 2-nitroethenyl benzene, 1-phenyl-2-nitroethene, beta-nitrosytrene, benzene, 2-nitroethenyl
5284459
[(E)-2-nitroethenyl]benzene
[O-][N+](=O)\C=C\C1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5153-67-3
C8H7NO2
149.15
MFCD00007402
PIAOLBVUVDXHHL-VOTSOKGWSA-N
trans-beta-nitrostyrene, 2-nitrovinyl benzene, e-2-nitrovinyl benzene, beta-nitrostyrene, 2-nitro-1-phenylethylene, e-beta-nitrostyrene, 2-nitroethenyl benzene, 1-phenyl-2-nitroethene, beta-nitrosytrene, benzene, 2-nitroethenyl
5284459
[(E)-2-nitroethenyl]benzene
[O-][N+](=O)\C=C\C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 56°C to 58°C
Color Yellow
Boiling Point 250°C to 260°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 149.15
Percent Purity 97+%
Physical Form Crystals
Chemical Name or Material trans-β-Nitrostyrene
Show More Show Less
compliance-icons
Product Identifier
  • trans-ß-Nitrostyrene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'