missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ trans-Dichlorobis(triethylphosphine)palladium(II)

Coupling of C-C bonds

Brand:  Alfa Aesar™ 039823.03

Code : NEW

Additional Details : CAS Number : 28425-04-9 Weight : 0.00100kg Transport : UN number : 3464 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 11309147

  • £124.97 / 1g


Estimated Shipment date
from Supplier 24-04-2018
Add to basket

Specifications

Chemical Identifiers

trans-Dichlorobis(triethylphosphine)palladium(II)
413.63
UN3464
28425-04-9
MFCD00191831

Specifications

trans-Dichlorobis(triethylphosphine)palladium(II)
413.63
UN3464
1g
28425-04-9
MFCD00191831
Soluble in chloroform, toluene and benzene
Hygroscopic
Videos
SDS