missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-Dichlorobis(diethylsulfide)platinum(II), Pt 43.7%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
It is used in Hydrosilylation. Platinum(II) complexes of diethylsulfide are convenient and versatile precursor for the preparation of a variety of organoplatinum(II) derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 1 g |
Solubility Information | Soluble in acetone,alcohol,ethyl acetate and benzene |
Formula Weight | 446.37 |
Concentration or Composition (by Analyte or Components) | Pt 43.7% |
Physical Form | Crystalline |
Chemical Name or Material | trans-Dichlorobis(diethylsulfide)platinum(II) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'