missing translation for 'onlineSavingsMsg'
Learn More

trans-Cinnamoyl chloride, 97%, Thermo Scientific Chemicals

Catalog Number p-7081848
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
17082-09-6
C9H7ClO
166.604
MFCD00000732
WOGITNXCNOTRLK-VOTSOKGWSA-N
cinnamoyl chloride, cinnamic acid chloride, cinnamic chloride, trans-3-phenylacryloyl chloride, 2-propenoyl chloride, 3-phenyl, cinnamoylchloride, e-cinnamoyl chloride, trans-cinnamoyl chloride, 2e-3-phenylprop-2-enoyl chloride, 3-phenyl-2-propenoyl chloride
5354261
(E)-3-phenylprop-2-enoyl chloride
C1=CC=C(C=C1)C=CC(=O)Cl
This item is not returnable. View return policy
17082-09-6
C9H7ClO
166.604
MFCD00000732
WOGITNXCNOTRLK-VOTSOKGWSA-N
cinnamoyl chloride, cinnamic acid chloride, cinnamic chloride, trans-3-phenylacryloyl chloride, 2-propenoyl chloride, 3-phenyl, cinnamoylchloride, e-cinnamoyl chloride, trans-cinnamoyl chloride, 2e-3-phenylprop-2-enoyl chloride, 3-phenyl-2-propenoyl chloride
5354261
(E)-3-phenylprop-2-enoyl chloride
C1=CC=C(C=C1)C=CC(=O)Cl

The derivative of this compound, trans-Cinnamoylimidazole is involved in the synthesis of trans-Cinnamoyl-Papain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The derivative of this compound, trans-Cinnamoylimidazole is involved in the synthesis of trans-Cinnamoyl-Papain.

Solubility
Reacts with water.

Notes
Store in cool place. Keep away from oxidizing agents. Incompatible with bases. Moisture sensitive.
TRUSTED_SUSTAINABILITY
Melting Point 35°C to 37°C
Density 1.162
Boiling Point 256°C to 258°C
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.5845
Quantity 100 g
UN Number UN3261
Beilstein 606265
Sensitivity Moisture sensitive
Merck Index 14,2301
Solubility Information Reacts with water.
Formula Weight 166.61
Percent Purity 97%
Chemical Name or Material trans-Cinnamoyl chloride
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'