missing translation for 'onlineSavingsMsg'
Learn More

trans-beta-Styrylboronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Catalog Number p-7047625
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy
83947-56-2
C14H19BO2
230.11
MFCD03453666
ARAINKADEARZLZ-ZHACJKMWSA-N
e-4,4,5,5-tetramethyl-2-styryl-1,3,2-dioxaborolane, e-phenylethenylboronic acid, pinacol ester, trans-styrylboronic acid, pinacol ester, beta-styrylboronic acid pinacol ester, trans-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl styrene, 4,4,5,5-tetramethyl-2-styryl-1,3,2-dioxaborolane, trans-beta-styrylboronic acid pinacol ester, trans-2-phenylvinylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-e-2-phenylethenyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-e-2-phenylvinyl-1,3,2-dioxaborolane
5708413
CC1(C)OB(OC1(C)C)\C=C\C1=CC=CC=C1
This item is not returnable. View return policy
83947-56-2
C14H19BO2
230.11
MFCD03453666
ARAINKADEARZLZ-ZHACJKMWSA-N
e-4,4,5,5-tetramethyl-2-styryl-1,3,2-dioxaborolane, e-phenylethenylboronic acid, pinacol ester, trans-styrylboronic acid, pinacol ester, beta-styrylboronic acid pinacol ester, trans-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl styrene, 4,4,5,5-tetramethyl-2-styryl-1,3,2-dioxaborolane, trans-beta-styrylboronic acid pinacol ester, trans-2-phenylvinylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-e-2-phenylethenyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-e-2-phenylvinyl-1,3,2-dioxaborolane
5708413
CC1(C)OB(OC1(C)C)\C=C\C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 27°C to 33°C
Boiling Point 128°C (4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.529
Quantity 250 mg
Beilstein 8617179
IUPAC Name 4,4,5,5-tetramethyl-2-[(E)-2-phenylethenyl]-1,3,2-dioxaborolane
Formula Weight 230.12
Percent Purity 99%
Chemical Name or Material trans-beta-Styrylboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'