missing translation for 'onlineSavingsMsg'
Learn More

(±)-trans-ACPD, Tocris Bioscience™

Group I and II mGlu agonist

Brand:  Tocris (Bio-Techne) 0187/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13447253

  • £189.63 / 10mg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Equimolecular mixture of (1S,3R)- and (1R,3S)-ACPD. Selective agonist for metabotropic glutamate receptors; active at both group I and group II mGlu receptors (EC50 values are 2, 15, 23 and ∽800 μM at mGluR2, mGluR1, mGluR5 and mGluR4 respectively).

(1S,3R)-ACPD and cis-ACPD also available.

Specifications

Chemical Identifiers

67684-64-4
173.168
1s,3s-1-aminocyclopentane-1,3-dicarboxylic acid, chembl29726, trans-acpd, cis-acpd, +/--trans-acpd, 1s, 3r-acpd, 1-amino-cis-cyclopentane-1,3-dicarboxylic acid, tocris-0186, tocris-0187, tocris-0284
(1S,3S)-1-aminocyclopentane-1,3-dicarboxylic acid
C7H11NO4
YFYNOWXBIBKGHB-FFWSUHOLSA-N
6604704
C1CC(CC1C(=O)O)(C(=O)O)N

Specifications

10mg
67684-64-4
1s,3s-1-aminocyclopentane-1,3-dicarboxylic acid, chembl29726, trans-acpd, cis-acpd, +/--trans-acpd, 1s, 3r-acpd, 1-amino-cis-cyclopentane-1,3-dicarboxylic acid, tocris-0186, tocris-0187, tocris-0284
C1CC(CC1C(=O)O)(C(=O)O)N
173.168
173.17
(±)-trans-ACPD
C7H11NO4
YFYNOWXBIBKGHB-FFWSUHOLSA-N
(1S,3S)-1-aminocyclopentane-1,3-dicarboxylic acid
6604704
>99%
Videos
Documents