missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Octene, 97%, Thermo Scientific Chemicals

Catalog Number p-7058745
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
14850-23-8
C8H16
112.216
MFCD00009476
IRUCBBFNLDIMIK-BQYQJAHWSA-N
trans-4-octene, e-4-octene, trans-n-4-octene, e-oct-4-ene, 4-octene, e, 4-octene, 4-octene, 4e, unii-fkz5737b1w, n-trans-4-octene, 4e-4-octene
5357253
(E)-oct-4-ene
CCCC=CCCC
This item is not returnable. View return policy
14850-23-8
C8H16
112.216
MFCD00009476
IRUCBBFNLDIMIK-BQYQJAHWSA-N
trans-4-octene, e-4-octene, trans-n-4-octene, e-oct-4-ene, 4-octene, e, 4-octene, 4-octene, 4e, unii-fkz5737b1w, n-trans-4-octene, 4e-4-octene
5357253
(E)-oct-4-ene
CCCC=CCCC

It is used in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.715 g/mL
Boiling Point 122°C to 123°C
Flash Point 21°C (70°F)
Odor Characteristic
Refractive Index 1.412
Quantity 2 g
UN Number UN3295
Beilstein 1719104
Solubility Information Sparingly soluble in water(10.7,mg/L).
Formula Weight 112.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material trans-4-Octene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'