missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Methoxycinnamaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number 10530532
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
24680-50-0
C10H10O2
162.19
MFCD00017343
AXCXHFKZHDEKTP-NSCUHMNNSA-N
4-methoxycinnamaldehyde, 3-4-methoxyphenyl acrylaldehyde, p-methoxycinnamaldehyde, e-3-4-methoxyphenyl acrylaldehyde, 3-4-methoxyphenyl-2-propenal, trans-4-methoxycinnamaldehyde, p-methoxy cinnamaldehyde, p-methoxycinnamic aldehyde, trans-p-methoxycinnamaldehyde, cinnamaldehyde, p-methoxy
641294
CHEBI:27952
COC1=CC=C(\C=C\C=O)C=C1
This item is not returnable. View return policy
24680-50-0
C10H10O2
162.19
MFCD00017343
AXCXHFKZHDEKTP-NSCUHMNNSA-N
4-methoxycinnamaldehyde, 3-4-methoxyphenyl acrylaldehyde, p-methoxycinnamaldehyde, e-3-4-methoxyphenyl acrylaldehyde, 3-4-methoxyphenyl-2-propenal, trans-4-methoxycinnamaldehyde, p-methoxy cinnamaldehyde, p-methoxycinnamic aldehyde, trans-p-methoxycinnamaldehyde, cinnamaldehyde, p-methoxy
641294
CHEBI:27952
COC1=CC=C(\C=C\C=O)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 55°C to 60°C
Color Orange-Yellow
Boiling Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble
IUPAC Name (E)-3-(4-methoxyphenyl)prop-2-enal
Formula Weight 162.19
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material trans-4-Methoxycinnamaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • trans-4-Methoxycinnamaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'