Learn More
trans-4-Hydroxycrotonic acid, Tocris Bioscience™
GHB receptor ligand
Brand:  Tocris (Bio-Techne) 053810
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13487693
  • £127.00 / 10mg
Estimated Shipment
02-07-2018
Add to basket
Description
Description
Binds to the γ-hydroxybutyric acid (GHB) receptor with higher affinity than GHB itself.? May be an endogenous ligand.
Specifications
Chemical Identifiers
trans-4-Hydroxycrotonic acid
C4H6O3
102.09
trans-4-Hydroxy-2-butenoic acid
Specifications
trans-4-Hydroxycrotonic acid
24587-49-3
C4H6O3
trans-4-Hydroxy-2-butenoic acid
10mg
>99%
102.09
Videos