missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Bromo-β-nitrostyrene, 99%, Thermo Scientific Chemicals

Catalog Number 11914591
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5153-71-9
C8H6BrNO2
228.05
LSGVHLGCJIBLMB-AATRIKPKSA-N
1-bromo-4-2-nitrovinyl benzene, e-1-bromo-4-2-nitrovinyl benzene, 1-bromo-4-e-2-nitroethenyl benzene, trans-4-bromo-beta-nitrostyrene, benzene, 1-bromo-4-1e-2-nitroethenyl, beta-nitro-4-bromostyrene, 4-bromo-.beta.-nitrostyrene, e-4-bromo-beta-nitrostyrene, 1-bromo-4-e-2-nitrovinyl benzene
688204
1-bromo-4-[(E)-2-nitroethenyl]benzene
C1=CC(=CC=C1C=C[N+](=O)[O-])Br
This item is not returnable. View return policy
5153-71-9
C8H6BrNO2
228.05
LSGVHLGCJIBLMB-AATRIKPKSA-N
1-bromo-4-2-nitrovinyl benzene, e-1-bromo-4-2-nitrovinyl benzene, 1-bromo-4-e-2-nitroethenyl benzene, trans-4-bromo-beta-nitrostyrene, benzene, 1-bromo-4-1e-2-nitroethenyl, beta-nitro-4-bromostyrene, 4-bromo-.beta.-nitrostyrene, e-4-bromo-beta-nitrostyrene, 1-bromo-4-e-2-nitrovinyl benzene
688204
1-bromo-4-[(E)-2-nitroethenyl]benzene
C1=CC(=CC=C1C=C[N+](=O)[O-])Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 148.0°C to 152.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 228.05
Percent Purity 99%
Physical Form Crystalline Powder or Powder
Chemical Name or Material trans-4-Bromo-β-nitrostyrene
compliance-icons
Product Identifier
  • trans-4-Bromo-beta-nitrostyrene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'