missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-4-(Boc-amino)cyclohexanecarboxylic acid, 98%, Thermo Scientific™
Click to view available options
Quantity:
0.25 g
500 mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Color | White |
Beilstein | 7641071 |
Formula Weight | 243.3 |
Percent Purity | 98% |
Quantity | 500 mg |
Physical Form | Powder |
Chemical Name or Material | trans-4-(Boc-amino)cyclohexanecarboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'