missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Boc-amino)cyclohexaneacetic acid, 97%, Thermo Scientific™

£20.60 - £299.00

Chemical Identifiers

CAS 189153-10-4
Molecular Formula C13H23NO4
Molecular Weight (g/mol) 257.33
MDL Number MFCD03844588
InChI Key IHXBNSUFUFFBRL-UHFFFAOYSA-N
Synonym 2-cis-4-tert-butoxycarbonyl amino cyclohexyl acetic acid, trans-n-boc-4-aminocyclohexyl acetic acid, 2-trans-4-tert-butoxycarbonyl amino cyclohexyl acetic acid, boc-1,4-trans-acha-oh, cis-4-boc-amino cyclohexyl acetic acid, trans-n-boc-4-aminocyclohexyl aceticacid, 4-tert-butoxycarbonylamino-cyclohexyl-acetic acid, trans-4-tert-butoxycarbonylamino cyclohexylacetic acid, 1r,4r-4-tert-butoxycarbonyl amino cyclohexyl acetic acid, 1s,4s-4-tert-butoxycarbonyl amino cyclohexyl acetic acid
PubChem CID 11253897
IUPAC Name 2-[4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexyl]acetic acid
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)CC(=O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15456976
SDS View Documents
Thermo Scientific Alfa Aesar
H62058.MD
250mg
£20.60
250mg
View alternative products
Save Up to 
»
Estimated Shipment: 18-11-2021
Please to purchase this item. Need a web account? Register with us today!
15498638
SDS View Documents
Thermo Scientific Alfa Aesar
H62058.03
1g
£60.10
1g
View alternative products
Save Up to 
»
Estimated Shipment: 18-11-2021
Please to purchase this item. Need a web account? Register with us today!
15408648
SDS View Documents
Thermo Scientific Alfa Aesar
H62058.06
5g
£299.00
5g
View alternative products
Save Up to 
»
Estimated Shipment: 18-11-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Applications
trans-4-(Boc-amino)cyclohexaneacetic acid used as a intermediate in pharmaceutical.

Solubility
Slightly soluble in water (1.1 g/L) (25°C)

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

189153-10-4
257.33
IHXBNSUFUFFBRL-UHFFFAOYSA-N
11253897
CC(C)(C)OC(=O)NC1CCC(CC1)CC(=O)O
C13H23NO4
MFCD03844588
2-cis-4-tert-butoxycarbonyl amino cyclohexyl acetic acid, trans-n-boc-4-aminocyclohexyl acetic acid, 2-trans-4-tert-butoxycarbonyl amino cyclohexyl acetic acid, boc-1,4-trans-acha-oh, cis-4-boc-amino cyclohexyl acetic acid, trans-n-boc-4-aminocyclohexyl aceticacid, 4-tert-butoxycarbonylamino-cyclohexyl-acetic acid, trans-4-tert-butoxycarbonylamino cyclohexylacetic acid, 1r,4r-4-tert-butoxycarbonyl amino cyclohexyl acetic acid, 1s,4s-4-tert-butoxycarbonyl amino cyclohexyl acetic acid
2-[4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexyl]acetic acid

Specifications

trans-4-(Boc-amino)cyclohexaneacetic acid
189153-10-4
MFCD03844588
Slightly soluble in water (1.1g/L) (25°C)
CC(C)(C)OC(=O)NC1CCC(CC1)CC(=O)O
257.33
257.32
250mg
C13H23NO4
2-cis-4-tert-butoxycarbonyl amino cyclohexyl acetic acid, trans-n-boc-4-aminocyclohexyl acetic acid, 2-trans-4-tert-butoxycarbonyl amino cyclohexyl acetic acid, boc-1,4-trans-acha-oh, cis-4-boc-amino cyclohexyl acetic acid, trans-n-boc-4-aminocyclohexyl aceticacid, 4-tert-butoxycarbonylamino-cyclohexyl-acetic acid, trans-4-tert-butoxycarbonylamino cyclohexylacetic acid, 1r,4r-4-tert-butoxycarbonyl amino cyclohexyl acetic acid, 1s,4s-4-tert-butoxycarbonyl amino cyclohexyl acetic acid
IHXBNSUFUFFBRL-UHFFFAOYSA-N
2-[4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexyl]acetic acid
11253897
97%
SDS
Product Certifications

RUO – Research Use Only