missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-4-(Boc-amino)cyclohexaneacetaldehyde, 97%, Thermo Scientific Chemicals
84.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
Description
trans-4-(Boc-amino)cyclohexaneacetaldehyde is an intermediate for the synthesis of cariprazine (C183490), a novel antipsychotic acting on dopamine D3/D2 receptors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
trans-4-(Boc-amino)cyclohexaneacetaldehyde is an intermediate for the synthesis of cariprazine (C183490), a novel antipsychotic acting on dopamine D3/D2 receptors.
Solubility
Sparingly Soluble (0.41 g/L) (25°C).
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
trans-4-(Boc-amino)cyclohexaneacetaldehyde is an intermediate for the synthesis of cariprazine (C183490), a novel antipsychotic acting on dopamine D3/D2 receptors.
Solubility
Sparingly Soluble (0.41 g/L) (25°C).
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Solubility Information | Sparingly Soluble (0.41g/L) (25°C). |
Formula Weight | 241.32 |
Percent Purity | 97% |
Quantity | 250 mg |
Chemical Name or Material | trans-4-(Boc-amino)cyclohexaneacetaldehyde |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'