missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Benzyloxy-N-Fmoc-L-proline, 98%, Thermo Scientific™

Catalog Number p-7055090
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
174800-02-3
C27H25NO5
443.499
MFCD00237023
XGFMHBUVVWZBFT-CLOONOSVSA-N
fmoc-hyp bzl-oh, 2s,4r-1-9h-fluoren-9-yl methoxy carbonyl-4-benzyloxy pyrrolidine-2-carboxylic acid, fmoc-o-benzyl-l-4-hydroxyproline, 2s,4r-4-benzyloxy-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carboxylic acid, ambotzfaa1790, fmoc-pro 4-obzl-oh, fmoc-hydroxyproline bzl-oh, fmoc-o-benzyl-l-4-trans-hydroxyproline, n-alpha-9-fluorenylmethoxycarbonyl-o-benzyl-l-4-hydroxyproline, 2s,4r-n-alpha-9-fluorenylmethyloxycarbonyl-4-oxybenzyl-l-proline
21063752
(2S,4R)-1-(9H-fluoren-9-ylmethoxycarbonyl)-4-phenylmethoxypyrrolidine-2-carboxylic acid
C1C(CN(C1C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)OCC5=CC=CC=C5
This item is not returnable. View return policy
174800-02-3
C27H25NO5
443.499
MFCD00237023
XGFMHBUVVWZBFT-CLOONOSVSA-N
fmoc-hyp bzl-oh, 2s,4r-1-9h-fluoren-9-yl methoxy carbonyl-4-benzyloxy pyrrolidine-2-carboxylic acid, fmoc-o-benzyl-l-4-hydroxyproline, 2s,4r-4-benzyloxy-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carboxylic acid, ambotzfaa1790, fmoc-pro 4-obzl-oh, fmoc-hydroxyproline bzl-oh, fmoc-o-benzyl-l-4-trans-hydroxyproline, n-alpha-9-fluorenylmethoxycarbonyl-o-benzyl-l-4-hydroxyproline, 2s,4r-n-alpha-9-fluorenylmethyloxycarbonyl-4-oxybenzyl-l-proline
21063752
(2S,4R)-1-(9H-fluoren-9-ylmethoxycarbonyl)-4-phenylmethoxypyrrolidine-2-carboxylic acid
C1C(CN(C1C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)OCC5=CC=CC=C5

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Formula Weight 443.49
Percent Purity 98%
Quantity 250 mg
Physical Form Powder
Chemical Name or Material trans-4-Benzyloxy-N-Fmoc-L-proline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'