missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ trans-4-Benzyloxy-N-Boc-L-proline dicyclohexylammonium salt, 95%

Brand:  Alfa Aesar™ H61561.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15456596

  • £13.36 / 1g


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

trans-4-Benzyloxy-N-Boc-L-proline dicyclohexylammonium salt
MFCD09260874
Boc-Hyp(Bzl)-OH.DCHA; (2S,4R)-4-Benzyloxy-1-Boc-pyrrolidine-2-carboxylic acid dicyclohexylammonium salt
502.69
5296858

Specifications

trans-4-Benzyloxy-N-Boc-L-proline dicyclohexylammonium salt
95%
MFCD09260874
Boc-Hyp(Bzl)-OH.DCHA; (2S,4R)-4-Benzyloxy-1-Boc-pyrrolidine-2-carboxylic acid dicyclohexylammonium salt
Solid
502.69
5296858
1g
Videos
SDS