missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Aminomethyl)cyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7025982
Click to view available options
Quantity:
10 g
50 g
250 g
1197-18-8
C8H15NO2
157.213
MFCD00001466
GYDJEQRTZSCIOI-UHFFFAOYSA-N
tranexamic acid, cyklokapron, tranexamsaeure, trans amcha, tranhexamic acid, trans-4-aminomethyl cyclohexanecarboxylic acid, transamin, amstat, tamcha, amikapron
5526
4-(aminomethyl)cyclohexane-1-carboxylic acid
C1CC(CCC1CN)C(=O)O
1197-18-8
C8H15NO2
157.213
MFCD00001466
GYDJEQRTZSCIOI-UHFFFAOYSA-N
tranexamic acid, cyklokapron, tranexamsaeure, trans amcha, tranhexamic acid, trans-4-aminomethyl cyclohexanecarboxylic acid, transamin, amstat, tamcha, amikapron
5526
4-(aminomethyl)cyclohexane-1-carboxylic acid
C1CC(CCC1CN)C(=O)O

It is an antifibrinolytic agent that is found to inhibit plasmin-induced fibrinolysis. Used in various hemorrhagic diseases , abnormal bleeding in operations. It is also used as a lysine analogue to characterize binding sites in plasminogen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an antifibrinolytic agent that is found to inhibit plasmin-induced fibrinolysis. Used in various hemorrhagic diseases , abnormal bleeding in operations. It is also used as a lysine analogue to characterize binding sites in plasminogen.

Solubility
Soluble in water (167 mg/ml), ethanol (<1mg/mL at 25°C). Very slightly soluble in ether, DMSO (<1mg/mL at 25°C), and alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point >300°C
Beilstein 2207452
Merck Index 14,9569
Solubility Information Soluble in water (167mg/ml),ethanol (<1mg/ml at 25°C). Very slightly soluble in ether,DMSO (<1mg/ml at 25°C),and alcohol.
Formula Weight 157.21
Percent Purity 97%
Quantity 10 g
Chemical Name or Material trans-4-(Aminomethyl)cyclohexanecarboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'