missing translation for 'onlineSavingsMsg'
Learn More

trans-4,5'-Dinitrodibenzo-18-crown-6, 98%, Acros Organics

Brand:  Acros Organics 357110010

Code : 29

Additional Details : CAS Number : 32082-45-4 Weight : 0.05100kg


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10088701

  • £51.60 / 1g


Estimated Shipment
16-03-2018
Add to basket


Specifications

Chemical Identifiers

trans-4, 5'-Dinitrodibenzo-18-crown-6
98%
C20H22N2O10
450.4

Specifications

trans-4, 5'-Dinitrodibenzo-18-crown-6
32082-45-4
98%
450.4
1g
Glass bottle
C20H22N2O10
Videos
SDS