missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ trans-4-(4-Vinylcyclohexyl)benzonitrile, 97%

£61.70 - £247.00

Chemical Identifiers

CAS 96184-42-8
Molecular Formula C15H17N
Molecular Weight (g/mol) 211.308
MDL Number MFCD11053490
InChI Key JHXJAWXZLWDDED-UHFFFAOYSA-N
Synonym 4-trans-4-vinylcyclohexyl benzonitrile, trans-4'-4-vinylcyclohexyl benzonitrile, trans-4-4-vinyl-cyclohexyl-benzonitrile, 4-4-ethenylcyclohexyl benzonitrile, 4-4-vinylcyclohexyl benzonitrile, jhxjawxzlwdded-mqmhxkeqsa-n, p-trans-4-vinylcyclohexyl benzonitrile, trans-4-4-vinylcyclohexyl benzonitrile, p-trans-4-vinylcyclohexyl-benzonitrile, 4-trans-4-ethenylcyclohexyl benzonitrile
PubChem CID 18724353
IUPAC Name 4-(4-ethenylcyclohexyl)benzonitrile
SMILES C=CC1CCC(CC1)C2=CC=C(C=C2)C#N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15491948
SDS View Documents
Alfa Aesar™
H55640.03
1g
£61.70
1g
View alternative products
Save Up to 
»
Estimated Shipment: 18-10-2021
Please to purchase this item. Need a web account? Register with us today!
15401958
SDS View Documents
Alfa Aesar™
H55640.06
5g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Chemical Identifiers

96184-42-8
211.308
JHXJAWXZLWDDED-UHFFFAOYSA-N
18724353
C=CC1CCC(CC1)C2=CC=C(C=C2)C#N
C15H17N
MFCD11053490
4-trans-4-vinylcyclohexyl benzonitrile, trans-4'-4-vinylcyclohexyl benzonitrile, trans-4-4-vinyl-cyclohexyl-benzonitrile, 4-4-ethenylcyclohexyl benzonitrile, 4-4-vinylcyclohexyl benzonitrile, jhxjawxzlwdded-mqmhxkeqsa-n, p-trans-4-vinylcyclohexyl benzonitrile, trans-4-4-vinylcyclohexyl benzonitrile, p-trans-4-vinylcyclohexyl-benzonitrile, 4-trans-4-ethenylcyclohexyl benzonitrile
4-(4-ethenylcyclohexyl)benzonitrile

Specifications

trans-4-(4-Vinylcyclohexyl)benzonitrile
1g
C15H17N
4-trans-4-vinylcyclohexyl benzonitrile, trans-4'-4-vinylcyclohexyl benzonitrile, trans-4-4-vinyl-cyclohexyl-benzonitrile, 4-4-ethenylcyclohexyl benzonitrile, 4-4-vinylcyclohexyl benzonitrile, jhxjawxzlwdded-mqmhxkeqsa-n, p-trans-4-vinylcyclohexyl benzonitrile, trans-4-4-vinylcyclohexyl benzonitrile, p-trans-4-vinylcyclohexyl-benzonitrile, 4-trans-4-ethenylcyclohexyl benzonitrile
C=CC1CCC(CC1)C2=CC=C(C=C2)C#N
211.308
211.31
57°C
96184-42-8
MFCD11053490
JHXJAWXZLWDDED-UHFFFAOYSA-N
4-(4-ethenylcyclohexyl)benzonitrile
18724353
97%
SDS
Product Certifications