missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(4-Vinylcyclohexyl)benzonitrile, 97%, Thermo Scientific™

Catalog Number 15491948
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
96184-42-8
C15H17N
211.308
MFCD11053490
JHXJAWXZLWDDED-UHFFFAOYSA-N
4-trans-4-vinylcyclohexyl benzonitrile, trans-4'-4-vinylcyclohexyl benzonitrile, trans-4-4-vinyl-cyclohexyl-benzonitrile, 4-4-ethenylcyclohexyl benzonitrile, 4-4-vinylcyclohexyl benzonitrile, jhxjawxzlwdded-mqmhxkeqsa-n, p-trans-4-vinylcyclohexyl benzonitrile, trans-4-4-vinylcyclohexyl benzonitrile, p-trans-4-vinylcyclohexyl-benzonitrile, 4-trans-4-ethenylcyclohexyl benzonitrile
18724353
4-(4-ethenylcyclohexyl)benzonitrile
C=CC1CCC(CC1)C2=CC=C(C=C2)C#N
This item is not returnable. View return policy
96184-42-8
C15H17N
211.308
MFCD11053490
JHXJAWXZLWDDED-UHFFFAOYSA-N
4-trans-4-vinylcyclohexyl benzonitrile, trans-4'-4-vinylcyclohexyl benzonitrile, trans-4-4-vinyl-cyclohexyl-benzonitrile, 4-4-ethenylcyclohexyl benzonitrile, 4-4-vinylcyclohexyl benzonitrile, jhxjawxzlwdded-mqmhxkeqsa-n, p-trans-4-vinylcyclohexyl benzonitrile, trans-4-4-vinylcyclohexyl benzonitrile, p-trans-4-vinylcyclohexyl-benzonitrile, 4-trans-4-ethenylcyclohexyl benzonitrile
18724353
4-(4-ethenylcyclohexyl)benzonitrile
C=CC1CCC(CC1)C2=CC=C(C=C2)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 57°C
Quantity 1 g
Formula Weight 211.31
Percent Purity 97%
Chemical Name or Material trans-4-(4-Vinylcyclohexyl)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'