missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-4-[4-(4-n-Pentylcyclohexyl)phenyl]benzonitrile, 99%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It finds its application as an intermediate of polymer-dispersed liquid crystals (PDLCs) are a relatively new class of materials that hold promise for many applications ranging from switchable windows to projection displays. These materials, which are simply a combined application of polymers and liquid crystals, are the focus of extensive research in the display industry.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It finds its application as an intermediate of polymer-dispersed liquid crystals (PDLCs) are a relatively new class of materials that hold promise for many applications ranging from switchable windows to projection displays. These materials, which are simply a combined application of polymers and liquid crystals, are the focus of extensive research in the display industry.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 95°C to 97°C |
Quantity | 1 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 331.5 |
Percent Purity | 99% |
Chemical Name or Material | trans-4-[4-(4-n-Pentylcyclohexyl)phenyl]benzonitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'