missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Hexene, 98%, Thermo Scientific Chemicals

Catalog Number p-4258993
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
13269-52-8
C6H12
84.16
ZQDPJFUHLCOCRG-AATRIKPKSA-N
trans-3-hexene, 3-hexene, e-3-hexene, 3-hexene, e, 3-hexene, 3e, unii-3co1lux1ar, e-hex-3-ene, 3co1lux1ar, alkenes, c6, cis-3-hexene
638066
(E)-hex-3-ene
CCC=CCC
This item is not returnable. View return policy
13269-52-8
C6H12
84.16
ZQDPJFUHLCOCRG-AATRIKPKSA-N
trans-3-hexene, 3-hexene, e-3-hexene, 3-hexene, e, 3-hexene, 3e, unii-3co1lux1ar, e-hex-3-ene, 3co1lux1ar, alkenes, c6, cis-3-hexene
638066
(E)-hex-3-ene
CCC=CCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -113.0°C
Density 0.6770g/mL
Boiling Point 67.0°C
Flash Point −12°C
Assay Percent Range 97.5% min
Packaging Glass bottle
Linear Formula C2H5CH=CHC2H5
Refractive Index 1.3935 to 1.3955
Quantity 1 g
Beilstein 01,II,192
Specific Gravity 0.677
Formula Weight 84.16
Percent Purity 98%
Chemical Name or Material trans-3-Hexene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • trans-3-Hexene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'