missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-3-Hexene, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Oligomers were prepared from the ozonolyses of cis- and trans-3-hexene and cis- and trans-2,5-dimethyl-3-hexene. Synthesis and characterization of aliphatic unsaturated polyesters is from trans-3-hexene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Oligomers were prepared from the ozonolyses of cis- and trans-3-hexene and cis- and trans-2,5-dimethyl-3-hexene. Synthesis and characterization of aliphatic unsaturated polyesters is from trans-3-hexene.
Solubility
Insoluble in water. Soluble in benzene, ligroin, ethanol, chloroform, ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Oligomers were prepared from the ozonolyses of cis- and trans-3-hexene and cis- and trans-2,5-dimethyl-3-hexene. Synthesis and characterization of aliphatic unsaturated polyesters is from trans-3-hexene.
Solubility
Insoluble in water. Soluble in benzene, ligroin, ethanol, chloroform, ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | -113°C |
Density | 0.677 |
Boiling Point | 66°C to 67°C |
Flash Point | −25°C (−13°F) |
Odor | Mild |
Refractive Index | 1.394 |
Quantity | 1 g |
UN Number | UN3295 |
Beilstein | 1718859 |
Solubility Information | Insoluble in water. Soluble in benzene,ligroin,ethanol,chloroform,ether. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'