missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ trans-3-Chloro-beta-nitrostyrene, 97%

Brand:  Alfa Aesar™ H64053.06

Code : NEW

Additional Details : CAS Number : 37888-03-2 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15481079

  • £54.70 / 5g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

trans-3-Chloro-beta-nitrostyrene
183.59
2087804
37888-03-2
MFCD00175518
trans-1-Chloro-3-(2-nitrovinyl)benzene

Specifications

trans-3-Chloro-beta-nitrostyrene
48° to 52°C
37888-03-2
97%
MFCD00175518
trans-1-Chloro-3-(2-nitrovinyl)benzene
Pale yellow
5g
Powder
183.59
2087804
Videos
SDS