missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Phenylcyclopropane-1-carboxylic acid, 97%, ACROS Organics™

Brand:  Acros Organics 130460050

Code : 29

Additional Details : CAS Number : 939-90-2 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 11352723

  • £53.50 / 5g


Estimated Shipment
22-06-2018
Add to basket


Specifications

Chemical Identifiers

trans-2-Phenylcyclopropane-1-carboxylic acid
97%
C6H5C3H4CO2H
MFCD00001292
97%
C10H10O2
162.19

Specifications

trans-2-Phenylcyclopropane-1-carboxylic acid
86.0° to 92.0°C
939-90-2
Glass bottle
97%
C10H10O2
162.19
Yellow or beige
5g
Authentic
Crystalline Powder
97%
C6H5C3H4CO2H
MFCD00001292
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • trans-2-Phenylcyclopropane-1-carboxylic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS