missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ trans-2-Octenoic acid, 94%

Brand:  Alfa Aesar™ L02058.09

Code : NEW

Additional Details : CAS Number : 1871-67-6 Weight : 0.01000kg Transport : UN number : 3265 Chem class : 8 Pack group : III

Quantity 10g

 View more versions of this product

Product Code. 11321497

  • £25.70 / 10g


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

trans-2-Octenoic acid
C8H14O2
MFCD00002706
1721115
1871-67-6
142.2
UN3265

Specifications

trans-2-Octenoic acid
94%
142.2
UN3265
101° to 102°C (5mmHg)
140°C (284°F)
1871-67-6
C8H14O2
MFCD00002706
1721115
10g
Characteristic
Videos
SDS