missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Octene 98%, Thermo Scientific™

Catalog Number 10205010
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
13389-42-9
C8H16
112.22
MFCD00009532
ILPBINAXDRFYPL-HWKANZROSA-N
trans-2-octene, 2-octene, e-2-octene, e-oct-2-ene, 2-octene, e, 2-octene, 2e, unii-g13g1yr8yw, oct-2-ene, g13g1yr8yw, octene-2
5364448
(2E)-oct-2-ene
CCCCC\C=C\C
This item is not returnable. View return policy
13389-42-9
C8H16
112.22
MFCD00009532
ILPBINAXDRFYPL-HWKANZROSA-N
trans-2-octene, 2-octene, e-2-octene, e-oct-2-ene, 2-octene, e, 2-octene, 2e, unii-g13g1yr8yw, oct-2-ene, g13g1yr8yw, octene-2
5364448
(2E)-oct-2-ene
CCCCC\C=C\C

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point -94.0°C
Color Colorless
Density 0.7180g/mL
Boiling Point 123.0°C
Flash Point 21°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)4CH=CHCH3
Refractive Index 1.4120 to 1.4140
Quantity 10 g
Specific Gravity 0.718
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether, petroleum and, coal-tar solvents
Formula Weight 112.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material trans-2-Octene, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • trans-2-Octene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P331-Do NOT induce vomiting.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'