missing translation for 'onlineSavingsMsg'
Learn More

trans-2-(4-chlorophenyl)vinylboronic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-4228545
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
154230-29-2
C8H8BClO2
182.41
MFCD02093767
HWSDRAPTZRYXHN-AATRIKPKSA-N
trans-2-4-chlorophenyl vinylboronic acid, 4-chlorostyryl boronic acid, e-2-4-chlorophenyl ethenyl boronic acid, e-4-chlorostyrylboronic acid, boronic acid, 1e-2-4-chlorophenyl ethenyl, e-2-4-chlorophenyl ethenylboronic acid, trans-2-4-chlorophenyl ethenylboronic acid, rarechem al ba 0169, e-2-4-chlorophenyl vinyl boronic acid, 4-chlorostyrylboronic acid
642694
[(E)-2-(4-chlorophenyl)ethenyl]boronic acid
OB(O)\C=C\C1=CC=C(Cl)C=C1
This item is not returnable. View return policy
154230-29-2
C8H8BClO2
182.41
MFCD02093767
HWSDRAPTZRYXHN-AATRIKPKSA-N
trans-2-4-chlorophenyl vinylboronic acid, 4-chlorostyryl boronic acid, e-2-4-chlorophenyl ethenyl boronic acid, e-4-chlorostyrylboronic acid, boronic acid, 1e-2-4-chlorophenyl ethenyl, e-2-4-chlorophenyl ethenylboronic acid, trans-2-4-chlorophenyl ethenylboronic acid, rarechem al ba 0169, e-2-4-chlorophenyl vinyl boronic acid, 4-chlorostyrylboronic acid
642694
[(E)-2-(4-chlorophenyl)ethenyl]boronic acid
OB(O)\C=C\C1=CC=C(Cl)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 133.0°C to 134.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 182.41
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material trans-2-(4-chlorophenyl)vinylboronic acid
compliance-icons
Product Identifier
  • trans-2-(4-Chlorophenyl)vinylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'