missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-(1R,2R)-(-)-N,N'-Dimethyl-1,2-cyclohexanediamine dihydrochloride, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Specifications
Assay Percent Range | 97.5 to max. 102.5 % min. (Argentometry) |
Packaging | Glass Bottle |
Quantity | 1 g |
Formula Weight | 215.17 |
Percent Purity | 98% |
Chemical Name or Material | trans-(1R,2R)-(-)-N,N'-Dimethyl-1,2-cyclohexanediamine dihydrochloride |
Safety and Handling
Product Identifier
- trans-(1R,2R)-(-)-N,N'-Dimethyl-1,2-cyclohexanediamine dihydrochloride, 98%
- Warning
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'