missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane is used to prepare C-2 hydroxyethyl imidazopyrrolo pyridines as JAK1 inhibitors. It is also used to prepare Mer kinase inhibitors for treatment of pediatric acute lymphoblastic leukemia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane is used to prepare C-2 hydroxyethyl imidazopyrrolo pyridines as JAK1 inhibitors. It is also used to prepare Mer kinase inhibitors for treatment of pediatric acute lymphoblastic leukemia.
Solubility
Slightly Soluble (4.4 g/L) (25°C).
Notes
Store in cool, dry place in tightly closed container. Keep container tightly closed. Store away from oxidizing agent.
trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane is used to prepare C-2 hydroxyethyl imidazopyrrolo pyridines as JAK1 inhibitors. It is also used to prepare Mer kinase inhibitors for treatment of pediatric acute lymphoblastic leukemia.
Solubility
Slightly Soluble (4.4 g/L) (25°C).
Notes
Store in cool, dry place in tightly closed container. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Solubility Information | Slightly Soluble (4.4g/L) (25°C). |
Formula Weight | 229.31 |
Percent Purity | 97% |
Quantity | 5 g |
Physical Form | Solid |
Chemical Name or Material | trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'