missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-1,2-Dibromocyclohexane, 99%, Thermo Scientific™
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Specifications
Color | Yellow |
Density | 1.7800g/mL |
Boiling Point | 145°C (100.0 mmHg) |
Flash Point | >110°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Linear Formula | C6H10Br2 |
Refractive Index | 1.5510 to 1.553 |
Quantity | 100 mL |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'