missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Dibenzoylethylene, 97%, Thermo Scientific Chemicals

Catalog Number 10666544
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
959-28-4
C16H12O2
236.27
MFCD00003083
WYCXGQSQHAXLPK-VAWYXSNFSA-N
trans-1,2-dibenzoylethylene, dibenzoylethylene, 1,2-dibenzoylethylene, 1,4-diphenyl-2-butene-1,4-dione, trans-1,4-diphenyl-2-butene-1,4-dione, 1,2-dibenzoylethene, ethylene, 1,2-dibenzoyl, usaf nd-57, 2-butene-1,4-dione, 1,4-diphenyl, trans-diphenacylidene
735960
(E)-1,4-diphenylbut-2-ene-1,4-dione
C1=CC=C(C=C1)C(=O)C=CC(=O)C2=CC=CC=C2
This item is not returnable. View return policy
959-28-4
C16H12O2
236.27
MFCD00003083
WYCXGQSQHAXLPK-VAWYXSNFSA-N
trans-1,2-dibenzoylethylene, dibenzoylethylene, 1,2-dibenzoylethylene, 1,4-diphenyl-2-butene-1,4-dione, trans-1,4-diphenyl-2-butene-1,4-dione, 1,2-dibenzoylethene, ethylene, 1,2-dibenzoyl, usaf nd-57, 2-butene-1,4-dione, 1,4-diphenyl, trans-diphenacylidene
735960
(E)-1,4-diphenylbut-2-ene-1,4-dione
C1=CC=C(C=C1)C(=O)C=CC(=O)C2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 108.0°C to 112.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5COCH=CHCOC6H5
Quantity 1 g
Beilstein 07, IV, 2578
Fieser 01,195
Solubility Information Solubility in water: insoluble
Formula Weight 236.27
Percent Purity 97%
Physical Form Crystals, Powder or Flakes
Chemical Name or Material trans-1, 2-Dibenzoylethylene, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'