missing translation for 'onlineSavingsMsg'
Learn More

(±)-trans-1,2-Diaminocyclohexane, 98%, Thermo Scientific Chemicals

Catalog Number p-2689130
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
1121-22-8
C6H14N2
114.19
MFCD00063747
SSJXIUAHEKJCMH-PHDIDXHHSA-N
1r,2r---1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, trans-1,2-diaminocyclohexane, trans-1,2-cyclohexanediamine, trans-cyclohexane-1,2-diamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r---1,2-cyclohexanediamine, 1r,2r-diaminocyclohexane, r-dach, 1r,2r-1,2-cyclohexanediamine
43806
(1R,2R)-cyclohexane-1,2-diamine
C1CCC(C(C1)N)N
This item is not returnable. View return policy
1121-22-8
C6H14N2
114.19
MFCD00063747
SSJXIUAHEKJCMH-PHDIDXHHSA-N
1r,2r---1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, trans-1,2-diaminocyclohexane, trans-1,2-cyclohexanediamine, trans-cyclohexane-1,2-diamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r---1,2-cyclohexanediamine, 1r,2r-diaminocyclohexane, r-dach, 1r,2r-1,2-cyclohexanediamine
43806
(1R,2R)-cyclohexane-1,2-diamine
C1CCC(C(C1)N)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 14.0°C to 15.0°C
Color Colorless to Yellow
Density 0.9510g/mL
Boiling Point 79.0°C to 81.0°C (15.0 mmHg)
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H10(NH2)2
Refractive Index 1.4885 to 1.4905
Quantity 10 mL
Specific Gravity 0.951
Solubility Information Solubility in water: soluble.
Formula Weight 114.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (±)-trans-1, 2-Diaminocyclohexane, 98%
Show More Show Less
compliance-icons
Product Identifier
  • (#1)-trans-1,2-Diaminocyclohexane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'