Learn More
Tranilast, Tocris Bioscience™
Antiallergic; inhibits inflammatory mediator release from mast cells
Brand:  Tocris (Bio-Techne) 109850
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13438483
  • £475.82 / 50mg
Estimated Shipment
01-07-2019
Description
Description
Antiallergic via inhibition of chemical mediator release from mast cells. Shown to be an effective inhibitor of angiogenesis. Demonstrated to antagonize the effects of angiotensin II on human arteries, possibly by an interaction at the level of the AT1 receptor. Inhibits TRPV2-mediated responses; binds to Aβ40 monomers and increases Aβ40 fibrillation.
Specifications
Chemical Identifiers
53902-12-8
327.336
tranilast, rizaben, trans-tranilast, tranilastum, n-3,4-dimethoxycinnamoyl anthranilic acid, tranilastum inn-latin, tranilast usan:inn:jan, tranpro, unii-hvf50smy6e, rizaben tn
CHEBI:77572
COC1=C(C=C(C=C1)C=CC(=O)NC2=CC=CC=C2C(=O)O)OC
C18H17NO5
NZHGWWWHIYHZNX-CSKARUKUSA-N
5282230
2-[[(E)-3-(3,4-dimethoxyphenyl)prop-2-enoyl]amino]benzoic acid
Specifications
Tranilast
53902-12-8
tranilast, rizaben, trans-tranilast, tranilastum, n-3,4-dimethoxycinnamoyl anthranilic acid, tranilastum inn-latin, tranilast usan:inn:jan, tranpro, unii-hvf50smy6e, rizaben tn
COC1=C(C=C(C=C1)C=CC(=O)NC2=CC=CC=C2C(=O)O)OC
327.336
CHEBI:77572
>99%
50mg
C18H17NO5
NZHGWWWHIYHZNX-CSKARUKUSA-N
2-[[(E)-3-(3,4-dimethoxyphenyl)prop-2-enoyl]amino]benzoic acid
5282230
327.34
Videos