missing translation for 'onlineSavingsMsg'
Learn More

TRAF-1, clone: 1F3, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-9791-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15557686

  • £77.04 / 25µg


Estimated Shipment date
from Supplier 01-06-2018
Add to basket

Description

Description

Description: The monoclonal antibody 1F3 recognizes human TRAF-1. TRAF-1 is a multifunctional adapter molecule and member of the TNF receptor associated factor protein family. TRAF proteins associate with various receptors of the TNF receptor superfamily to mediate signal transduction. TRAF-1 plays a role in regulating cell survival and apoptosis, regulates the signaling of NF kappa B and JNK, and forms a heterodimeric complex with TRAF-2. TRAF-1 also interacts with cell surface receptors such as CD30, CD40, and CD357 (AITR), and cytoplasmic proteins like NIK, RIP2, A20, and TRIP. Expression of TRAF-1 can be induced by Epstein-Barr Virus (EBV) and TNF alpha.Applications Reported: This 1F3 antibody has been reported for use in western blotting, microscopy, immunohistochemical staining, and immunocytochemistry.Applications Tested: This 1F3 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded human tissue using low pH antigen retrieval and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

TRAF-1
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q13077
Epstein-Bar virus-induced protein 6
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
1F3
Unconjugated
TRAF1
Purified
TRAF1
IgG2a, kappa
25μg
Store at 2-8°C.
Primary
7185
Videos

For Research Use Only.