missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TraceGOLD TG-ALC1 GC Columns

Analyze blood alcohols with this ultra-low bleed column for enhanced sensitivity and reproducibility

Brand:  Thermo Scientific™ 26074-3960

Code : A0

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 13446846

  • £456.00 / Each


Estimated Shipment date
from Supplier 16-04-2020
Add to basket

Description

Description

Redefining Your Expectations for GC:
  • Low bleed delivers excellent signal-to-noise ratio, sensitivity, and mass spectral integrity.
  • Fast analysis time facilitates full separation of blood alcohols.
  • Highly inert phase ensures excellent peak shape and sensitivity.
  • Applications
  • Blood alcohol analysis
  • Abused inhalant anesthetics
  • γ-hydroxybutyrate (GHB)
  • γ-butyrolactone (GBL)
  • Glycols
  • Common industrial solvents
A Leap Forward in Performance with Low Bleed and Superior Inertness:
  • A low-bleed, stable column produces consistent results.
  • Low baseline noise enables improved limits of detection with enhanced resolution of low level analytes.
  • Minimizing column bleed extends column lifetime, leading to higher productivity.
  • Reducing column bleed increases reproducibility of the stationary phase over time, resulting in greater run-to-run and batch-to-batch reproducibility.
  • Superior inertness ensures excellent peak shape and sensitivity, especially for highly active or difficult-to-analyze compounds.
  • Proprietary phase facilitates analysis of difficult-to-analyze compounds
Similar To:
  • Rtx™ BAC Plus 1
  • Rtx™ BAC Plus 2
Specifications

Specifications

0.53 mm
TraceGOLD™ TG-ALC1
30 m
GC/MS
260°C
Videos
Documents